Gene Transcripts
- Gene Symbol
- Mrc1
- Entrez Gene ID
- 17533
- Gene Name
- mannose receptor, C type 1
- Gene Aliases
- AW259686, CD206, MR
- Chromosome Location
- Chr. 2: 14229414 - 14332023 on Build GRCm38
- UniGene ID
- Mm.2019
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_008625.2 |
NP_032651.2
|
30 |
4678 |
|
GenBank mRNA |
Z11974.1 |
- |
30 |
4678 |
|
View Full Product Details