Gene Transcripts

Gene Symbol
Insl5
Entrez Gene ID
23919
Gene Name
insulin-like 5
Gene Aliases
RIF2
Chromosome Location
Chr. 4: 103017872 - 103026842 on Build GRCm38
UniGene ID
Mm.100163
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290648.1 NP_001277577.1 3 556
NM_011831.2 NP_035961.1 2 562
GenBank mRNA AF076971.1 - 2 558
AF133817.1 - 2 565
BC010968.1 - 2 557 4217383

View Full Product Details