Gene Transcripts

Gene Symbol
Cul1
Entrez Gene ID
26965
Gene Name
cullin 1
Gene Aliases
Chromosome Location
Chr. 6: 47452273 - 47526139 on Build GRCm38
UniGene ID
Mm.87611
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012042.3 NP_036172.1 4 783
GenBank mRNA AF083216.1 - 3 507
AF136441.1 - 3 638
AF176910.1 - 4 672
AK147004.1 - 4 765
AK164274.1 - 4 750
AK165978.1 - 4 723
AK208612.1 - 2 276
BC004836.1 - 3 592
BC029260.1 - 4 783 2812400

View Full Product Details