Gene Transcripts
- Gene Symbol
- Cul1
- Entrez Gene ID
- 26965
- Gene Name
- cullin 1
- Gene Aliases
- Chromosome Location
- Chr. 6: 47452273 - 47526139 on Build GRCm38
- UniGene ID
- Mm.87611
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012042.3 |
NP_036172.1
|
5, 6 |
863 |
|
GenBank mRNA |
AF083216.1 |
- |
4, 5 |
587 |
|
AF136441.1 |
- |
4, 5 |
718 |
|
AF176910.1 |
- |
5, 6 |
752 |
|
AK147004.1 |
- |
5, 6 |
845 |
|
AK164274.1 |
- |
5, 6 |
830 |
|
AK165978.1 |
- |
5, 6 |
804 |
|
AK208612.1 |
- |
3, 4 |
356 |
|
BC004836.1 |
- |
4, 5 |
672 |
|
BC029260.1 |
- |
5, 6 |
863 |
2812400 |
View Full Product Details