Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001139520.1 | NP_001132992.1 | 2 | 393 | |
NM_018851.3 | NP_061339.3 | 2 | 393 | ||
GenBank mRNA | AK054490.1 | - | 2 | 292 | |
AK078936.1 | - | 2 | 336 | ||
AK128954.1 | - | 2 | 264 | ||
AK149865.1 | - | 2 | 268 | ||
AK150964.1 | - | 2 | 108 | ||
AK151335.1 | - | 2 | 267 | ||
AK152815.1 | - | 2 | 108 | ||
AK153390.1 | - | 2 | 267 | ||
AK170688.1 | - | 2 | 292 | ||
AK171565.1 | - | 2 | 292 | ||
BC012721.1 | - | 2 | 330 | 4037046 | |
BC067198.1 | - | 2 | 316 | 6826682 | |
U15635.1 | - | 2 | 317 |