Gene Transcripts

Gene Symbol
CHRNA10
Entrez Gene ID
57053
Gene Name
cholinergic receptor nicotinic alpha 10 subunit
Gene Aliases
Chromosome Location
Chr. 11: 3665587 - 3673208 on Build GRCh38
UniGene ID
Hs.732806
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303034.1 NP_001289963.1 5 1731
NM_001303035.1 NP_001289964.1 5 1664
NM_020402.3 NP_065135.2 5 1669
XM_011520234.1 XP_011518536.1 4 4865
GenBank mRNA AF199235.2 - 5 1669
AJ278118.1 - 5 1639

View Full Product Details