Gene Transcripts

Gene Symbol
Ubxn6
Entrez Gene ID
66530
Gene Name
UBX domain protein 6
Gene Aliases
1200008L11Rik, 2210415J11Rik, AU040909, Ubxd1, Ubxdc2
Chromosome Location
Chr. 17: 56068249 - 56074989 on Build GRCm38
UniGene ID
Mm.28000
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024432.2 NP_077752.1 2, 3 325
GenBank mRNA AF272895.1 - 2, 3 340
AK008942.1 - 2, 3 334
AK009117.1 - 1, 2 107
AK075705.1 - 2, 3 353
AK082510.1 - 364
AK148170.1 - 293
AK161273.1 - 2, 3 346
BC016412.1 - 2, 3 254 4215883
BC049963.1 - 2, 3 325 2655935

View Full Product Details