Gene Transcripts

Gene Symbol
Ubxn6
Entrez Gene ID
66530
Gene Name
UBX domain protein 6
Gene Aliases
1200008L11Rik, 2210415J11Rik, AU040909, Ubxd1, Ubxdc2
Chromosome Location
Chr. 17: 56068249 - 56074989 on Build GRCm38
UniGene ID
Mm.28000
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024432.2 NP_077752.1 7 715
GenBank mRNA AF272895.1 - 7 730
AK008942.1 - 7 724
AK009117.1 - 6 497
AK075705.1 - 5 1779
AK082510.1 - 754
AK131187.1 - 1 2487
AK148170.1 - 683
AK161273.1 - 7 736
BC016412.1 - 7 644 4215883
BC049963.1 - 7 715 2655935

View Full Product Details