Gene Transcripts

Gene Symbol
Asrgl1
Entrez Gene ID
66514
Gene Name
asparaginase like 1
Gene Aliases
2410004D18Rik, ALP, ALP1, AU040643, AW060375
Chromosome Location
Chr. 19: 9111719 - 9135566 on Build GRCm38
UniGene ID
Mm.272847
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025610.3 NP_079886.2 4 470
GenBank mRNA AK006147.1 - 4 506
AK010381.1 - 4 512
AK030209.1 - 4 471
BC016106.1 - 2 222 4921265

View Full Product Details