Gene Transcripts

Gene Symbol
EPB42
Entrez Gene ID
2038
Gene Name
erythrocyte membrane protein band 4.2
Gene Aliases
PA, SPH5
Chromosome Location
Chr. 15: 43197227 - 43225856 on Build GRCh38
UniGene ID
Hs.368642
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000119.2 NP_000110.2 11 2138
NM_001114134.1 NP_001107606.1 11 2048
XM_005254225.1 XP_005254282.1 10 1780
XM_011521349.2 XP_011519651.1 12 2866
XM_011521350.2 XP_011519652.1 13 2598
XM_011521351.2 XP_011519653.1 13 2460
XM_011521352.2 XP_011519654.1 11 2041
XM_011521353.2 XP_011519655.1 10 1971
XM_011521354.2 XP_011519656.1 10 1452
GenBank mRNA AK297146.1 - 11 1822
AK297663.1 - 10 1698
BC096093.4 - 11 1827 40001236
BC096094.2 - 11 1827 40001240
BC096095.4 - 6 1262
BC099627.1 - 10 1611 40001239
M29399.1 - 11 1935
M30646.1 - 11 1975
M30647.1 - 11 2065
M60298.1 - 11 2297

View Full Product Details