Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001167921.1 | NP_001161393.1 | 2 | 210 | |
NM_001167922.1 | NP_001161394.1 | 2 | 312 | ||
NM_026584.3 | NP_080860.3 | 2 | 198 | ||
GenBank mRNA | AK007492.1 | - | 1 | 155 | |
AK021183.1 | - | 2 | 163 | ||
AK088342.1 | - | 2 | 223 | ||
AK146803.1 | - | 2 | 185 | ||
AK150415.1 | - | 2 | 156 | ||
AK153370.1 | - | 2 | 156 | ||
AK158783.1 | - | 2 | 292 | ||
BC020016.1 | - | 2 | 178 | 3501113 | |
BX516160.1 | - | 2 | 449 | ||
BY256665.1 | - | 2 | 199 | ||
CX239577.1 | - | 2 | 210 |