Gene Transcripts

Gene Symbol
Apitd1
Entrez Gene ID
69928
Gene Name
apoptosis-inducing, TAF9-like domain 1
Gene Aliases
2610040C18Rik, 2810407L01Rik, CENP-S
Chromosome Location
Chr. 4: 149128349 - 149137600 on Build GRCm38
UniGene ID
Mm.260893
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027263.2 NP_081539.1 2 178
GenBank mRNA AK011725.1 - 2 127
AK012995.1 - 2 160
AK013038.1 - 2 160
BC048521.1 - 2 155 6741262
BC138926.1 - 2 119 8861948
BC138927.1 - 2 119 8861949
DV656255.1 - 2 178

View Full Product Details