Gene Transcripts
- Gene Symbol
- Fyttd1
- Entrez Gene ID
- 69823
- Gene Name
- forty-two-three domain containing 1
- Gene Aliases
- 2010005M05Rik, 3830411L18Rik, 3830421F13Rik, 4933423A17Rik, AI098218, AW539567, C85190
- Chromosome Location
- Chr. 16: 32877581 - 32908963 on Build GRCm38
- UniGene ID
- Mm.12831
- Species
- Mus musculus
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AK031662.1 |
- |
|
2076 |
|
Chr. - - [-] |
View Full Product Details