Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001141950.1 | NP_001135422.1 | 6 | 698 | |
NM_027797.3 | NP_082073.1 | 6 | 767 | ||
GenBank mRNA | AK017460.1 | - | 6 | 768 | |
AK049405.1 | - | 2 | 628 | ||
AK134821.1 | - | 6 | 699 | ||
AK169083.1 | - | 6 | 657 | ||
BC022756.1 | - | 6 | 642 | 4922223 |