Gene Transcripts
- Gene Symbol
- Mms19
- Entrez Gene ID
- 72199
- Gene Name
- MMS19 (MET18 S. cerevisiae)
- Gene Aliases
- 2410001K24Rik, 2610042O15Rik, AI316855, C79368, C86341, Mms19l
- Chromosome Location
- Chr. 19: 41943707 - 41981159 on Build GRCm38
- UniGene ID
- Mm.218940
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_028152.3 |
NP_082428.1
|
3, 4 |
292 |
|
GenBank mRNA |
AF319949.1 |
- |
3, 4 |
286 |
|
AK010326.1 |
- |
3, 4 |
296 |
|
AK011754.1 |
- |
3, 4 |
286 |
|
AK028317.1 |
- |
3, 4 |
292 |
|
AK030619.1 |
- |
3, 4 |
293 |
|
AK036983.1 |
- |
3, 4 |
286 |
|
AK078688.1 |
- |
3, 4 |
287 |
|
AK087633.1 |
- |
3, 4 |
293 |
|
BC018614.1 |
- |
3, 4 |
270 |
|
BC050817.1 |
- |
3, 4 |
272 |
6332214 |
View Full Product Details