Gene Transcripts

Gene Symbol
Brdt
Entrez Gene ID
114642
Gene Name
bromodomain, testis-specific
Gene Aliases
7420412D09Rik, Brd6, Fsrg3
Chromosome Location
Chr. 5: 107331091 - 107387061 on Build GRCm38
UniGene ID
Mm.182836
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_054054.2 NP_473395.2 8 1308
GenBank mRNA AF358660.1 - 8 1343

View Full Product Details