Gene Transcripts

Gene Symbol
Snx4
Entrez Gene ID
69150
Gene Name
sorting nexin 4
Gene Aliases
1810036H14Rik, AI037066
Chromosome Location
Chr. 16: 33251456 - 33299562 on Build GRCm38
UniGene ID
Mm.28196
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080557.2 NP_542124.1 4, 5 562
GenBank mRNA AK007706.1 - 4, 5 604
AK030657.1 - 4, 5 575
AK031368.1 - 574
AK135356.1 - 4, 5 569
BC006043.1 - 4, 5 552
BC016599.1 - 4, 5 562 2644991

View Full Product Details