Gene Transcripts

Gene Symbol
Olfml3
Entrez Gene ID
99543
Gene Name
olfactomedin-like 3
Gene Aliases
2810002E22Rik, AI115209, AW550633, HNOEL-iso, ONT3, mONT3
Chromosome Location
Chr. 3: 103735394 - 103738001 on Build GRCm38
UniGene ID
Mm.211535
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133859.2 NP_598620.2 3 1416
GenBank mRNA AK002518.1 - 3 1378
AK076084.1 - 3 1382
AK131879.1 - 1380
AK143728.1 - 1968
BC005485.1 - 3 1363 3485412

View Full Product Details