Gene Transcripts
- Gene Symbol
- Usp39
- Entrez Gene ID
- 28035
- Gene Name
- ubiquitin specific peptidase 39
- Gene Aliases
- AA408960, AI894154, CGI-21, D6Wsu157e, SAD1
- Chromosome Location
- Chr. 6: 72318676 - 72345217 on Build GRCm38
- UniGene ID
- Mm.281900
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_138592.4 |
NP_613058.1
|
13 |
2192 |
|
GenBank mRNA |
AK018595.1 |
- |
13 |
2194 |
|
View Full Product Details