Gene Transcripts

Gene Symbol
Ubtd1
Entrez Gene ID
226122
Gene Name
ubiquitin domain containing 1
Gene Aliases
BC016129
Chromosome Location
Chr. 19: 41981763 - 42034650 on Build GRCm38
UniGene ID
Mm.22948
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145500.3 NP_663475.1 3 792
GenBank mRNA AK078479.1 - 3 792
AK216640.1 - 1 16
BC016129.1 - 3 618 4920477

View Full Product Details