Gene Transcripts

Gene Symbol
Fbxo21
Entrez Gene ID
231670
Gene Name
F-box protein 21
Gene Aliases
2810425J22Rik, AU016673, mKIAA0875
Chromosome Location
Chr. 5: 117976735 - 118010201 on Build GRCm38
UniGene ID
Mm.21912
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145564.3 NP_663539.1 9, 10 1362
GenBank mRNA AK049723.1 - 9, 10 1363
BC021871.1 - 9, 10 1399 3709413

View Full Product Details