Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145605.2 | NP_663580.2 | 3 | 344 | |
GenBank mRNA | AK089954.1 | - | 3 | 307 | |
AK161674.1 | - | 3 | 320 | ||
BC012312.1 | - | 3 | 344 | 2631632 | |
BC023738.1 | - | 3 | 315 | 5352770 | |
BY086869.1 | - | 3 | 330 |