Gene Transcripts

Gene Symbol
Rhot2
Entrez Gene ID
214952
Gene Name
ras homolog gene family, member T2
Gene Aliases
Arht2, Miro2
Chromosome Location
Chr. 17: 25838838 - 25844851 on Build GRCm38
UniGene ID
Mm.40907
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145999.2 NP_666111.1 4 618
GenBank mRNA AK142183.1 - 3 314
AY311390.1 - 4 618
BC029777.1 - 4 251 4950061
BC034062.1 - 4 273 4207654

View Full Product Details