Gene Transcripts

Gene Symbol
Olfr139
Entrez Gene ID
259005
Gene Name
olfactory receptor 139
Gene Aliases
M5, MOR255-2
Chromosome Location
Chr. 11: 74044325 - 74045272 on Build GRCm38
UniGene ID
Mm.377784
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_147003.1 NP_667214.1 1 833
GenBank mRNA BC104101.2 - 1 833
BC104102.1 - 1 833 40043111

View Full Product Details