Gene Transcripts

Gene Symbol
Olfr623
Entrez Gene ID
259126
Gene Name
olfactory receptor 623
Gene Aliases
MOR1-4
Chromosome Location
Chr. 7: 103660244 - 103661318 on Build GRCm38
UniGene ID
Mm.377863
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_147122.2 NP_667333.2 1 381
GenBank mRNA BC119275.1 - 1 381

View Full Product Details