Gene Transcripts

Gene Symbol
Slco4a1
Entrez Gene ID
108115
Gene Name
solute carrier organic anion transporter family, member 4a1
Gene Aliases
OATP-E, Slc21a12
Chromosome Location
Chr. 2: 180456250 - 180474853 on Build GRCm38
UniGene ID
Mm.133687
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_148933.1 NP_683735.1 12 2646
GenBank mRNA AK033598.1 - 12 2466
AK172688.1 - 12 2396
BC026987.1 - 3 518
BC030719.1 - 12 2646 4501966
BC030720.1 - 12 2595 4505508
BC033602.1 - 3 1635

View Full Product Details