Gene Transcripts

Gene Symbol
Rbks
Entrez Gene ID
71336
Gene Name
ribokinase
Gene Aliases
5230400M11Rik, AI326806, RBSK
Chromosome Location
Chr. 5: 31624439 - 31697698 on Build GRCm38
UniGene ID
Mm.22519
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153196.1 NP_694876.1 2, 3 242
GenBank mRNA AK017229.1 - 2, 3 260
AK138939.1 - 2, 3 260
BC023339.1 - 2, 3 242 5253827

View Full Product Details