Gene Transcripts
- Gene Symbol
- RPS16
- Entrez Gene ID
- 6217
- Gene Name
- ribosomal protein S16
- Gene Aliases
- S16
- Chromosome Location
- Chr. 19: 39433137 - 39436020 on Build GRCh38
- UniGene ID
- Hs.397609
- Species
- Homo sapiens
- Gene Symbol
- ZNF90
- Entrez Gene ID
- 7643
- Gene Name
- zinc finger protein 90
- Gene Aliases
- HTF9
- Chromosome Location
- Chr. 19: 20077994 - 20127032 on Build GRCh38
- UniGene ID
- Hs.55304
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK298173.1 |
- |
3 |
596 |
|
View Full Product Details