Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001102607.1 | NP_001096077.1 | 8 | 3480 | |
NM_172927.3 | NP_766515.2 | 8 | 3515 | ||
GenBank mRNA | AK054356.1 | - | 8 | 3516 | |
AK087828.1 | - | 8 | 3556 | ||
AM748260.1 | - | 6 | 3200 | ||
AM748261.1 | - | 1 | 296 | ||
BC150695.1 | - | 7 | 3299 | 9087606 |