Gene Transcripts

Gene Symbol
Casc4
Entrez Gene ID
319996
Gene Name
cancer susceptibility candidate 4
Gene Aliases
D130060C09Rik, VGFG2573
Chromosome Location
Chr. 2: 121866970 - 121936220 on Build GRCm38
UniGene ID
Mm.292109
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001205369.1 NP_001192298.1 1, 2 561
NM_001205370.1 NP_001192299.1 1, 2 561
NM_177054.5 NP_796028.2 1, 2 561
NM_199038.3 NP_950239.2 1, 2 561
GenBank mRNA AK051613.1 - 1, 2 403
AK080364.1 - 1, 2 412
AK160708.1 - 1, 2 402
AY484583.1 - 1, 2 401
AY484585.1 - 1, 2 401
BC034556.1 - 1, 2 424
BC064446.1 - 1, 2 561 6843743

View Full Product Details