Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001415.3 | NP_001406.1 | 4 | 384 | |
GenBank mRNA | AB451230.1 | - | 4 | 363 | |
AB451353.1 | - | 4 | 363 | ||
AK222577.1 | - | 4 | 376 | ||
AK290397.1 | - | 4 | 376 | ||
AK312247.1 | - | 4 | 376 | ||
BC019906.1 | - | 4 | 384 | 4419438 | |
L19161.1 | - | 4 | 363 |