Gene Transcripts

Gene Symbol
CCND1
Entrez Gene ID
595
Gene Name
cyclin D1
Gene Aliases
BCL1, D11S287E, PRAD1, U21B31
Chromosome Location
Chr. 11: 69641105 - 69654474 on Build GRCh38
UniGene ID
Hs.523852
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053056.2 NP_444284.1 3 735
GenBank mRNA AK313136.1 - 3 684
BC000076.2 - 3 669 3508088
BC001501.2 - 3 669 3507598
BC014078.2 - 3 669 4124540
BC023620.2 - 3 662 4650919
BC025302.1 - 3 671 5457963
BT019844.1 - 3 526
BT019845.1 - 3 526
CR536538.1 - 3 526
CR542099.1 - 3 526
M64349.1 - 3 670
M73554.1 - 3 667
M74092.1 - 3 675
X59798.1 - 3 673

View Full Product Details