Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004929.3 | NP_004920.1 | 5 | 528 | |
GenBank mRNA | AK054881.1 | - | 5 | 526 | |
AK295585.1 | - | 4 | 247 | ||
AK312491.1 | - | 5 | 526 | ||
AK315959.1 | - | 4 | 234 | ||
BC006478.1 | - | 5 | 516 | 2822391 | |
BC020864.1 | - | 5 | 526 | 4592497 | |
X06661.1 | - | 5 | 367 |