Gene Transcripts

Gene Symbol
IFIT3
Entrez Gene ID
3437
Gene Name
interferon induced protein with tetratricopeptide repeats 3
Gene Aliases
CIG-49, GARG-49, IFI60, IFIT4, IRG2, ISG60, P60, RIG-G, cig41
Chromosome Location
Chr. 10: 89327819 - 89340968 on Build GRCh38
UniGene ID
Hs.47338
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031683.3 NP_001026853.1 2 1240
NM_001289758.1 NP_001276687.1 3 1469
NM_001289759.1 NP_001276688.1 3 1362
NM_001549.5 NP_001540.2 2 1351
GenBank mRNA AF026939.1 - 2 1190
AF083470.1 - 2 1174
AK290427.1 - 2 1261
AK297137.1 - 3 1160
AK312675.1 - 2 1171
BC001383.2 - 2 1173 3050764
BC004977.1 - 2 1146 2906188
BT007284.1 - 1 1096
U52513.1 - 2 1160

View Full Product Details