Gene Transcripts

Gene Symbol
CXCL10
Entrez Gene ID
3627
Gene Name
C-X-C motif chemokine ligand 10
Gene Aliases
C7, IFI10, INP10, IP-10, SCYB10, crg-2, gIP-10, mob-1
Chromosome Location
Chr. 4: 76021116 - 76023536 on Build GRCh38
UniGene ID
Hs.632586
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001565.3 NP_001556.2 3 344
GenBank mRNA BC010954.1 - 3 309
X02530.1 - 3 305

View Full Product Details