Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001114380.1 | NP_001107852.1 | 7 | 817 | |
NM_002209.2 | NP_002200.2 | 9 | 1066 | ||
XM_005255313.1 | XP_005255370.1 | 9 | 986 | ||
XM_006721044.1 | XP_006721107.1 | 7 | 768 | ||
XM_011545849.1 | XP_011544151.1 | 9 | 999 | ||
GenBank mRNA | AK314246.1 | - | 9 | 1020 | |
BC008777.2 | - | 7 | 750 | 3142951 | |
Y00796.1 | - | 9 | 978 |