Gene Transcripts

Gene Symbol
SLC5A3
Entrez Gene ID
6526
Gene Name
solute carrier family 5 member 3
Gene Aliases
BCW2, SMIT, SMIT1, SMIT2
Chromosome Location
Chr. 21: 34073523 - 34106262 on Build GRCh38
UniGene ID
Hs.302742
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006933.6 NP_008864.4 2 1230
GenBank mRNA AX750578.1 - 2 1183

View Full Product Details