Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001324095.1 | NP_001311024.1 | 8 | 937 | |
NM_006438.4 | NP_006429.2 | 6 | 706 | ||
XM_005250756.3 | XP_005250813.1 | 6 | 607 | ||
GenBank mRNA | AB002631.1 | - | 6 | 614 | |
AK291791.1 | - | 6 | 706 | ||
AY359038.1 | - | 6 | 630 | ||
BC103815.1 | - | 6 | 650 | 40000432 | |
BC103816.1 | - | 6 | 650 | 40000433 |