Gene Transcripts

Gene Symbol
DRAP1
Entrez Gene ID
10589
Gene Name
DR1 associated protein 1
Gene Aliases
NC2-alpha
Chromosome Location
Chr. 11: 65919257 - 65921577 on Build GRCh38
UniGene ID
Hs.356742
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006442.3 NP_006433.2 5 612
GenBank mRNA BC010025.2 - 5 420
BG719065.1 - 5 615
BQ773919.1 - 3 407
X96506.1 - 5 367

View Full Product Details