Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006691.3 | NP_006682.2 | 2 | 475 | |
GenBank mRNA | AF118108.1 | - | 2 | 279 | |
AF127670.2 | - | 2 | 309 | ||
AK075443.1 | - | 2 | 389 | ||
AK312462.1 | - | 2 | 473 | ||
AY358925.1 | - | 2 | 348 | ||
BC026231.1 | - | 2 | 234 | 4712863 | |
DA924658.1 | - | 2 | 475 |