Gene Transcripts
- Gene Symbol
- BLOC1S6
- Entrez Gene ID
- 26258
- Gene Name
- biogenesis of lysosomal organelles complex 1 subunit 6
- Gene Aliases
- BLOS6, HPS9, PA, PALLID, PLDN
- Chromosome Location
- Chr. 15: 45587123 - 45609716 on Build GRCh38
- UniGene ID
- Hs.7037
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001311255.1 |
NP_001298184.1
|
2, 3 |
381 |
|
NM_012388.3 |
NP_036520.1
|
2, 3 |
547 |
|
NR_132356.1 |
|
2, 3 |
249 |
|
NR_132357.1 |
|
2, 3 |
249 |
|
GenBank mRNA |
AF080470.1 |
- |
2, 3 |
451 |
|
AK315581.1 |
- |
2, 3 |
265 |
|
AL596952.1 |
- |
2, 3 |
320 |
|
BC004819.1 |
- |
2, 3 |
268 |
3450397 |
DA859617.1 |
- |
2, 3 |
197 |
|
DB289156.1 |
- |
2, 3 |
192 |
|
View Full Product Details