Gene Transcripts

Gene Symbol
ANKRD11
Entrez Gene ID
29123
Gene Name
ankyrin repeat domain 11
Gene Aliases
ANCO-1, ANCO1, LZ16, T13
Chromosome Location
Chr. 16: 89267619 - 89490561 on Build GRCh38
UniGene ID
Hs.335003
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256182.1 NP_001243111.1 10 5271
NM_001256183.1 NP_001243112.1 9 5197
NM_013275.5 NP_037407.4 9 5200
XM_006721181.1 XP_006721244.1 9 4977
XM_006721184.2 XP_006721247.1 8 5015
XM_011523051.2 XP_011521353.1 10 5260
XM_011523053.2 XP_011521355.1 10 5366
XM_011523054.2 XP_011521356.1 10 5280
XM_011523055.2 XP_011521357.1 11 5565
XM_011523056.2 XP_011521358.1 10 5302
XM_011523057.2 XP_011521359.1 9 5183
XM_017023182.1 XP_016878671.1 9 5633
XM_017023183.1 XP_016878672.1 10 5369
XM_017023184.1 XP_016878673.1 10 5366
XM_017023185.1 XP_016878674.1 8 5007
XM_017023186.1 XP_016878675.1 12 5743
XM_017023187.1 XP_016878676.1 8 5095
GenBank mRNA AY373756.1 - 9 5203
AY533563.1 - 10 5028

View Full Product Details