Gene Transcripts

Gene Symbol
ANAPC4
Entrez Gene ID
29945
Gene Name
anaphase promoting complex subunit 4
Gene Aliases
APC4
Chromosome Location
Chr. 4: 25377226 - 25418498 on Build GRCh38
UniGene ID
Hs.152173
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286756.1 NP_001273685.1 29 2510
NM_013367.2 NP_037499.2 29 2507
XM_005248159.2 XP_005248216.1 16 1404
XM_011513838.1 XP_011512140.1 29 2387
XM_017008138.1 XP_016863627.1 29 2436
GenBank mRNA AF191338.1 - 28 2378
AK023562.1 - 2 2165
AK023597.1 - 26 2121
AK292336.1 - 29 2427
AL353932.1 - 8 3642
BC059383.1 - 29 2480 30349524
CA312572.1 - 1 179

View Full Product Details