Gene Transcripts

Gene Symbol
ZMYND10
Entrez Gene ID
51364
Gene Name
zinc finger MYND-type containing 10
Gene Aliases
BLU, CILD22, FLU
Chromosome Location
Chr. 3: 50341106 - 50346028 on Build GRCh38
UniGene ID
Hs.526735
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308379.1 NP_001295308.1 4 503
NM_015896.3 NP_056980.2 4 503
XM_005265216.2 XP_005265273.1 3 676
GenBank mRNA AB209621.1 - 1 2928
AK058135.1 - 4 433
AK096525.1 - 4 1611
AK223343.1 - 4 418
AK308086.1 - 4 402
AK315123.1 - 4 489
AL833828.1 - 4 432
BC033732.1 - 4 399 5168611
DB036846.1 - 4 503
U70824.1 - 4 457
U70880.1 - 4 457

View Full Product Details