Gene Transcripts

Gene Symbol
ST6GALNAC1
Entrez Gene ID
55808
Gene Name
ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 1
Gene Aliases
HSY11339, SIAT7A, ST6GalNAcI, STYI
Chromosome Location
Chr. 17: 76617769 - 76643838 on Build GRCh38
UniGene ID
Hs.105352
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289107.1 NP_001276036.1 4 1298
NM_018414.4 NP_060884.1 3 1161
NR_110309.1 3 1161
XM_011524995.2 XP_011523297.1 3 1081
XM_011524996.2 XP_011523298.1 3 1081
XM_017024842.1 XP_016880331.1 3 1081
XM_017024843.1 XP_016880332.1 3 1081
XM_017024844.1 XP_016880333.1 4 1181
GenBank mRNA AK000113.1 - 2 315
AK310906.1 - 3 1097
AY096001.1 - 3 961
AY358918.1 - 3 1106
BC022462.1 - 3 1016 4816713
Y11339.2 - 3 1021

View Full Product Details