Gene Transcripts
- Gene Symbol
- HEATR6
- Entrez Gene ID
- 63897
- Gene Name
- HEAT repeat containing 6
- Gene Aliases
- ABC1
- Chromosome Location
- Chr. 17: 60041363 - 60078931 on Build GRCh38
- UniGene ID
- Hs.221899
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_022070.4 |
NP_071353.4
|
18, 19 |
2776 |
|
XM_005257570.3 |
XP_005257627.1
|
19, 20 |
2964 |
|
XM_011525110.2 |
XP_011523412.1
|
17, 18 |
2663 |
|
XM_011525111.2 |
XP_011523413.1
|
17, 18 |
2625 |
|
XM_017024936.1 |
XP_016880425.1
|
18, 19 |
3037 |
|
XM_017024937.1 |
XP_016880426.1
|
18, 19 |
4713 |
|
XM_017024938.1 |
XP_016880427.1
|
18, 19 |
2769 |
|
XM_017024939.1 |
XP_016880428.1
|
18, 19 |
3040 |
|
GenBank mRNA |
AF349752.1 |
- |
19, 20 |
2721 |
|
AK025740.1 |
- |
11, 12 |
1843 |
|
AK026070.1 |
- |
11, 12 |
1616 |
|
AK127708.1 |
- |
18, 19 |
2770 |
|
AK314655.1 |
- |
18, 19 |
2779 |
|
BC119771.1 |
- |
16, 17 |
2430 |
40108435 |
BC119813.1 |
- |
18, 19 |
2767 |
40108426 |
BX640819.1 |
- |
18, 19 |
2767 |
|
BX640831.1 |
- |
18, 19 |
2767 |
|
CR627418.1 |
- |
18, 19 |
2766 |
|
View Full Product Details