Gene Transcripts
- Gene Symbol
- PGBD1
- Entrez Gene ID
- 84547
- Gene Name
- piggyBac transposable element derived 1
- Gene Aliases
- HUCEP-4, SCAND4, dJ874C20.4
- Chromosome Location
- Chr. 6: 28281537 - 28303681 on Build GRCh38
- UniGene ID
- Hs.144527
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001184743.1 |
NP_001171672.1
|
5, 6 |
1209 |
|
NM_032507.3 |
NP_115896.1
|
5, 6 |
1189 |
|
XM_006715228.2 |
XP_006715291.1
|
4, 5 |
859 |
|
XM_011514942.2 |
XP_011513244.1
|
4, 5 |
840 |
|
XM_011514943.2 |
XP_011513245.1
|
4, 5 |
825 |
|
XM_017011361.1 |
XP_016866850.1
|
4, 5 |
866 |
|
XM_017011362.1 |
XP_016866851.1
|
5, 6 |
975 |
|
GenBank mRNA |
AK223446.1 |
- |
5, 6 |
1174 |
|
AK291228.1 |
- |
5, 6 |
1154 |
|
BC032507.1 |
- |
5, 6 |
1139 |
|
BC069033.1 |
- |
5, 6 |
1035 |
|
BC128585.1 |
- |
5, 6 |
1036 |
40128523 |
D88259.1 |
- |
4, 5 |
769 |
|
View Full Product Details