Gene Transcripts

Gene Symbol
C2orf73
Entrez Gene ID
129852
Gene Name
chromosome 2 open reading frame 73
Gene Aliases
Chromosome Location
Chr. 2: 54329961 - 54383673 on Build GRCh38
UniGene ID
Hs.732626
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100396.1 NP_001093866.1 5 674
XM_005264133.1 XP_005264190.1 4 466
XM_005264134.3 XP_005264191.1 4 549
XM_011532510.2 XP_011530812.1 5 744
XM_011532513.2 XP_011530815.1 4 647
XM_017003323.1 XP_016858812.1 5 569
XM_017003324.1 XP_016858813.1 4 632
GenBank mRNA AK097617.1 - 4 466
BC031669.1 - 5 637
BC126244.1 - 4 435 8991960
BC126246.1 - 4 435 8991962
BC144191.1 - 3 319
BC144192.1 - 5 469 9052712

View Full Product Details