Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001114938.2 | NP_001108410.2 | 5 | 841 | |
NM_001190182.1 | NP_001177111.1 | 5 | 814 | ||
XM_011540812.1 | XP_011539114.1 | 4 | 1668 | ||
XM_011540814.1 | XP_011539116.1 | 4 | 1668 | ||
XM_011540820.1 | XP_011539122.1 | 4 | 1668 | ||
XM_017000450.1 | XP_016855939.1 | 4 | 1641 | ||
XM_017000451.1 | XP_016855940.1 | 3 | 1572 | ||
XM_017000452.1 | XP_016855941.1 | 5 | 1569 | ||
XM_017000453.1 | XP_016855942.1 | 4 | 1668 | ||
XM_017000454.1 | XP_016855943.1 | 4 | 739 | ||
XM_017000455.1 | XP_016855944.1 | 3 | 286 | ||
XM_017000456.1 | XP_016855945.1 | 3 | 287 | ||
XM_017000457.1 | XP_016855946.1 | 4 | 1668 | ||
XM_017000458.1 | XP_016855947.1 | 4 | 1668 | ||
GenBank mRNA | AK057646.1 | - | 4 | 744 | |
AK298104.1 | - | 5 | 812 | ||
AK303939.1 | - | 5 | 839 | ||
AK310879.1 | - | 2 | 1167 | ||
BC029888.1 | - | 3 | 464 | 5175485 | |
BC128177.1 | - | 3 | 430 | 40113154 |