Gene Transcripts

Gene Symbol
ZNF550
Entrez Gene ID
162972
Gene Name
zinc finger protein 550
Gene Aliases
Chromosome Location
Chr. 19: 57541836 - 57561512 on Build GRCh38
UniGene ID
Hs.180257
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001277090.1 NP_001264019.1 4 829
NM_001277091.1 NP_001264020.1 4 829
NM_001277092.1 NP_001264021.1 4 829
XM_011526567.2 XP_011524869.1 4 826
XM_011526568.2 XP_011524870.1 4 1919
XM_017026401.1 XP_016881890.1 4 826
GenBank mRNA AK122867.1 - 4 830
AK312802.1 - 5 781
AL833214.1 - 1 398
BC034810.1 - 4 805 4823435
BC053858.1 - 4 803 6044705
BX538032.1 - 4 816

View Full Product Details